missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha Glucosidase 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35393-100ul
This item is not returnable.
View return policy
Description
alpha Glucosidase 2 Polyclonal antibody specifically detects alpha Glucosidase 2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| alpha Glucosidase 2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| Alpha-glucosidase 2, EC 3.2.1, EC 3.2.1.84, G2ANGLUII, Glucosidase II subunit alpha, glucosidase, alpha; neutral AB, GluII, KIAA0088alpha glucosidase II alpha subunit, neutral alpha-glucosidase AB | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human alpha Glucosidase 2 (NP_001265123.1).,, Sequence:, MTRFRIDELEPRRPRYRVPDVLVADPPIARLSVSGRDENSVELTMAEGPYKIILTARPFRLDLLEDRSLLLSVNARGLLEFEHQRAPRVSQGSKDPAEGDGAQPEETPRDGDKPEETQGKAEKDEPGAWEETFKTHSDSKPYGPMSVGLDFSLPGMEHVYGIPEHADNLRLKVTEGGEPYRLYNLDVFQYELYNPMALYGSVPVLLAHNPHRDLGIFWLNAAETWVDISSNTAGKTLFGKMMDYLQGSGETPQTDVRWMS | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 23193 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction