Learn More
alpha GalCer:CD1d Complex Monoclonal Antibody (L363), PE, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 12-2019-82
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: This L363 monoclonal antibody reacts with alpha-galactosylceramide bound to the mouse MHC class I-like protein CD1d. This complex is recognized by invariant natural killer T (iNKT) cells, a subset of T lymphocytes that expresses the invariant TCR alpha chain Valpha14Jalpha18. Activation of iNKT cells leads to production of Th1 and Th2-associated cytokines as well as dendritic cell maturation. The L363 antibody has been reported to neutralize iNKT cell activation by alpha-galactosylceramide:CD1d complexes. Applications Reported: This L363 antibody has been reported for use in flow cytometric analysis. Applications Tested: This L363 antibody has been tested by flow cytometric analysis of alpha-galactosylceramide-loaded mouse CD1d-transfected cells. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
This complex is recognized by invariant natural killer T (iNKT) cells, a subset of T lymphocytes that expresses the invariant TCR alpha chain Va14Ja18. Activation of iNKT cells leads to production of Th1 and Th2-associated cytokines as well as dendritic cell maturation.Specifications
alpha GalCer:CD1d Complex | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P11609, Q9JHE4 | |
Cd1d1, Gal3st1 | |
Affinity chromatography | |
RUO | |
12479, 53897 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
L363 | |
PE | |
Cd1d1 | |
3'-phosphoadenosine-5'-phosphosulfate:Galcer sulfotransferase; 3'-phosphoadenylylsulfate:galactosylceramide 3'-sulfotransferase; 3'-phosphoadenylylsulfate-galactosylceramide 3'-sulfotransferase; AI747460; Antigen-presenting glycoprotein CD1d1; CD1.1; Cd1a; Cd1d; CD1d.1; Cd1d1; CD1d1 antigen; cerebroside sulfotransferase; Cst; Gal3st1; galactose-3-O-sulfotransferase 1; galactosylceramide sulfotransferase; GalCer sulfotransferase; Gcst; Ly-38; T-cell surface glycoprotein CD1d1 | |
Mouse | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
For Research Use Only.