Learn More
alpha GalCer:CD1d Complex Monoclonal Antibody (L363), Functional Grade, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£7770.00 - £12950.00
Specifications
| Antigen | alpha GalCer:CD1d Complex |
|---|---|
| Clone | L363 |
| Concentration | 1 mg/mL |
| Applications | Flow Cytometry, Functional Assay |
| Classification | Monoclonal |
Description
This L363 monoclonal antibody reacts with alpha-galactosylceramide bound to the mouse MHC class I-like protein CD1d. This complex is recognized by invariant natural killer T (iNKT) cells, a subset of T lymphocytes that expresses the invariant TCR alpha chain Valpha14Jalpha18. Activation of iNKT cells leads to production of Th1 and Th2-associated cytokines as well as dendritic cell maturation. The L363 antibody has been reported to neutralize iNKT cell activation by alpha-galactosylceramide:CD1d complexes. Applications Reported: This L363 antibody has been reported for use in functional assays. Applications Tested: This L363 antibody has been tested by flow cytometric analysis of alpha-galactosylcermide-loaded mouse CD1d-transfected cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/ μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.
alpha GalCer:CD1d Complex Monoclonal antibody specifically detects alpha GalCer:CD1d Complex in Mouse samples. It is validated for Flow Cytometry, Functional AssaySpecifications
| alpha GalCer:CD1d Complex | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.2 | |
| AI747460; Antigen-presenting glycoprotein CD1d1; Cd1.1; Cd1a; Cd1d; CD1d.1; Cd1d1; CD1d1 antigen; Ly-38; T-cell surface glycoprotein CD1d1 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
| L363 | |
| Flow Cytometry, Functional Assay | |
| Functional Grade | |
| Mouse | |
| Mouse | |
| P11609, Q9JHE4 | |
| 12479, 53897 | |
| Primary | |
| 4° C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.