Learn More
alpha GalCer:CD1d Complex Monoclonal Antibody (L363), Functional Grade, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£138.00
Specifications
Antigen | alpha GalCer:CD1d Complex |
---|---|
Clone | L363 |
Concentration | 1 mg/mL |
Applications | Flow Cytometry, Functional Assay |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15208597
|
Affymetrix eBioscience
16-2019-82 |
100 μg |
£138.00
100µg |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: This L363 monoclonal antibody reacts with alpha-galactosylceramide bound to the mouse MHC class I-like protein CD1d. This complex is recognized by invariant natural killer T (iNKT) cells, a subset of T lymphocytes that expresses the invariant TCR alpha chain Valpha14Jalpha18. Activation of iNKT cells leads to production of Th1 and Th2-associated cytokines as well as dendritic cell maturation. The L363 antibody has been reported to neutralize iNKT cell activation by alpha-galactosylceramide:CD1d complexes. Applications Reported: This L363 antibody has been reported for use in functional assays. Applications Tested: This L363 antibody has been tested by flow cytometric analysis of alpha-galactosylcermide-loaded mouse CD1d-transfected cells. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.
This complex is recognized by invariant natural killer T (iNKT) cells, a subset of T lymphocytes that expresses the invariant TCR alpha chain Va14Ja18. Activation of iNKT cells leads to production of Th1 and Th2-associated cytokines as well as dendritic cell maturation.Specifications
alpha GalCer:CD1d Complex | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative; pH 7.2 | |
3'-phosphoadenosine-5'-phosphosulfate:Galcer sulfotransferase; 3'-phosphoadenylylsulfate:galactosylceramide 3'-sulfotransferase; 3'-phosphoadenylylsulfate-galactosylceramide 3'-sulfotransferase; AI747460; Antigen-presenting glycoprotein CD1d1; CD1.1; Cd1a; Cd1d; CD1d.1; Cd1d1; CD1d1 antigen; cerebroside sulfotransferase; Cst; Gal3st1; galactose-3-O-sulfotransferase 1; galactosylceramide sulfotransferase; GalCer sulfotransferase; Gcst; Ly-38; T-cell surface glycoprotein CD1d1 | |
Cd1d1, Gal3st1 | |
Primary | |
4° C | |
Cd1d1 |
L363 | |
Flow Cytometry, Functional Assay | |
Functional Grade | |
Mouse | |
Mouse | |
P11609, Q9JHE4 | |
12479, 53897 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.