missing translation for 'onlineSavingsMsg'
Learn More

Alpha-Galactosyl-C18-ceramide, TRC

Product Code. 30400953
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30400953 10 mg
30400843 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30400953 Supplier Toronto Research Chemicals Supplier No. TRCG18495010MG
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 148347-40-4
Molecular Formula C42H81NO8
Molecular Weight (g/mol) 728.09
IUPAC Name N-[(E,2S,3R)-3-hydroxy-1-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoctadec-4-en-2-yl]octadecanamide
SMILES CCCCCCCCCCCCCCCCCC(=O)N[C@@H](CO[C@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O)[C@H](O)\C=C\CCCCCCCCCCCCC

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C42H81NO8/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-38(46)43-35(34-50-42-41(49)40(48)39(47)37(33-44)51-42)36(45)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2/h29,31,35-37,39-42,44-45,47-49H,3-28,30,32-34H2,1-2H3,(H,43,46)/b31-29+/t35-,36+,37+,39-,40-,41+,42-/m0/s1
Formula Weight 727.5962
Chemical Name or Material a-Galactosyl-C18-ceramide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.