missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ alpha Galactosidase Monoclonal Antibody (C8)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA541589
This item is not returnable.
View return policy
Description
alpha Galactosidase Monoclonal Antibody for Western Blot, IHC (P)
This gene encodes a homodimeric glycoprotein that hydrolyses the terminal alpha-galactosyl moieties from glycolipids and glycoproteins. This enzyme predominantly hydrolyzes ceramide trihexoside, and it can catalyze the hydrolysis of melibiose into galactose and glucose. A variety of mutations in this gene affect the synthesis, processing, and stability of this enzyme, which causes Fabry disease, a rare lysosomal storage disorder that results from a failure to catabolize alpha-D-galactosyl glycolipid moieties.
Specifications
| alpha Galactosidase | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P06280 | |
| GLA | |
| Recombinant protein Galactosidase Alpha. The antigen corresponds to amino acid range 81-429 of the target protein. | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| Immunohistochemistry (Paraffin), Western Blot | |
| C8 | |
| Unconjugated | |
| GLA | |
| Agalsidase; agalsidase alfa; Ags; Alpha-D-galactosidase A; alpha-D-galactoside galactohydrolase; alpha-D-galactoside galactohydrolase 1; alpha-gal A; alpha-galactosidase A; GALA; galactosidase alpha; galactosidase, alpha; GLA; Melibiase | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 2717 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction