missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha galactosidase A Mouse anti-Human, Clone: 2B2C5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66121-1-IG-20UL
This item is not returnable.
View return policy
Description
This gene encodes a homodimeric glycoprotein that hydrolyses the terminal alpha-galactosyl moieties from glycolipids and glycoproteins. This enzyme predomitly hydrolyzes ceramide trihexoside, and it can catalyze the hydrolysis of melibiose into galactose and glucose. A variety of mutations in this gene affect the synthesis, processing, and stability of this enzyme, which causes Fabry disease, a rare lysosomal storage disorder that results from a failure to catabolize alpha-D-galactosyl glycolipid moieties.Specifications
| Alpha galactosidase A | |
| Monoclonal | |
| 2.39 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P06280 | |
| GLA | |
| Alpha galactosidase A Fusion Protein Ag7505 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 2B2C5 | |
| Unconjugated | |
| GLA | |
| Alpha galactosidase A, galactosidase, alpha | |
| Mouse | |
| Protein A | |
| RUO | |
| 2717 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction