missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Galactosidase A/GLA Rabbit anti-Human, Clone: 1, Novus Biologicals™
Rabbit Monoclonal Antibody
£328.00 - £409.00
Specifications
| Antigen | alpha-Galactosidase A/GLA |
|---|---|
| Clone | 1 |
| Dilution | Western Blot 1:1000-1:5000, ELISA 1:5000-1:10000, Immunoprecipitation 4-6 ul/mg of lysate |
| Applications | Western Blot, ELISA, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18251638
|
Novus Biologicals
NBP2-90047-50UL |
50 μg |
£328.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18238396
|
Novus Biologicals
NBP2-90047-100UL |
100 μg |
£409.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
alpha-Galactosidase A/GLA Monoclonal specifically detects alpha-Galactosidase A/GLA in Human samples. It is validated for ELISA, Immunoprecipitation.Specifications
| alpha-Galactosidase A/GLA | |
| Western Blot 1:1000-1:5000, ELISA 1:5000-1:10000, Immunoprecipitation 4-6 ul/mg of lysate | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 2717 | |
| IgG | |
| Protein A purified |
| 1 | |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Agalsidase, agalsidase alfa, Alpha-D-galactosidase A, Alpha-D-galactoside galactohydrolase, alpha-D-galactoside galactohydrolase 1, alpha-gal A, alpha-galactosidase A, EC 3.2.1, EC 3.2.1.22, GALA, galactosidase, alpha, melibiase | |
| Recombinant Human alpha-Galactosidase A/GLA protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title