missing translation for 'onlineSavingsMsg'
Learn More

Alpha Fodrin Antibody (SPTAN1/3505), DyLight 594, Novus Biologicals™

Product Code. 18375474 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18375474 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18375474 Supplier Novus Biologicals Supplier No. NBP314184DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Alpha Fodrin Monoclonal antibody specifically detects Alpha Fodrin in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Alpha Fodrin
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SPTAN1/3505
Conjugate DyLight 594
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Alpha-II spectrin, EIEE5, FLJ17738, FLJ44613, Fodrin alpha chain, NEAS, spectrin alpha chain, brain, spectrin, alpha, non-erythrocytic 1 (alpha-fodrin), Spectrin, non-erythroid alpha chain, SPTA2
Host Species Mouse
Immunogen Recombinant fragment of human Alpha Fodrin protein (around aa2351-2475) (exact sequence is proprietary)) (Uniprot: Q13813)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6709
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.