missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ alpha-Fetoprotein/AFP Native Protein

Product Code. 18689037 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 18689037

Brand: Novus Biologicals™ NBP262398

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Native Protein

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mg/mL
For Use With (Application) Immunoassay
Formulation TBS
Gene ID (Entrez) 174
Molecular Weight (g/mol) MolecularWeight-theroretical: 63 kDa
Quantity 1 mg
Storage Requirements Store at 4°C.
Gene Alias Alpha-1-fetoprotein, Alpha-fetoglobulin, alpha-fetoprotein, FETA, HP, HPAFP
Gene Symbol AFP
Research Category Cancer, Cancer Stem Cells, Developmental Biology, Stem Cell Markers, Tumor Biomarkers
Purity or Quality Grade >95%, by SDS-PAGE
Protein alpha-Fetoprotein/AFP
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.