missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Fetoprotein/AFP Antibody (189508), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB13692-SP
This item is not returnable.
View return policy
Description
alpha-Fetoprotein/AFP Monoclonal specifically detects alpha-Fetoprotein/AFP in Human samples. It is validated for Western Blot, Simple Western, Knockout Validated.
Specifications
| alpha-Fetoprotein/AFP | |
| Monoclonal | |
| Unconjugated | |
| PBS with No Preservative | |
| AFP | |
| Human umbilical cord serum-derived alpha -Fetoprotein/AFP | |
| 25 μg | |
| Cancer, Developmental Biology | |
| 174 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, KnockDown | |
| 189508 | |
| Western Blot 0.1 - 0.5 ug/ml, Simple Western, Knockout Validated 0.05 ug/ml | |
| Alpha-1-fetoprotein, Alpha-fetoglobulin, alpha-fetoprotein, FETA, HP, HPAFP | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction