missing translation for 'onlineSavingsMsg'
Learn More

Alpha-Ethyl-Alpha-(1-demethylethyl)-Verapamil Hydrochloride, TRC

Product Code. 30380907
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380907 10 mg -
30374696 25 mg -
30383069 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380907 Supplier Toronto Research Chemicals Supplier No. TRCE91310010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 190850-49-8
Molecular Formula C26H36N2O4.HCl
Molecular Weight (g/mol) 477.04
Synonym (2RS)-2-(3,4-Dimethoxyphenyl)-5-[[2-(3,4-dimethoxyphenyl)-ethyl](methyl)amino]-2-ethylpentanenitrile Hydrochloride, Verapamil Hydrochloride Imp. H (EP) as Hydrochloride
IUPAC Name 2-(3,4-dimethoxyphenyl)-5-[2-(3,4-dimethoxyphenyl)ethyl-methylamino]-2-ethylpentanenitrile;hydrochloride
SMILES Cl.CCC(CCCN(C)CCc1ccc(OC)c(OC)c1)(C#N)c2ccc(OC)c(OC)c2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C26H36N2O4.ClH/c1-7-26(19-27,21-10-12-23(30-4)25(18-21)32-6)14-8-15-28(2)16-13-20-9-11-22(29-3)24(17-20)31-5;/h9-12,17-18H,7-8,13-16H2,1-6H3;1H
Formula Weight 476.2442
Chemical Name or Material (2RS)-2-(3,4-Dimethoxyphenyl)-5-[(2-(3,4-dimethoxyphenyl)-ethyl)(methyl)amino]-2-ethylpentanenitrile Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.