missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha Endosulfine Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00273-100ul
This item is not returnable.
View return policy
Description
alpha Endosulfine Polyclonal antibody specifically detects alpha Endosulfine in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
| alpha Endosulfine | |
| Polyclonal | |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 4-6 uL/mg of lysate | |
| alpha-endosulfine, ARPP-19eMGC4319, endosulfine alpha, MGC78563, MGC8394 | |
| Produced in rabbits immunized with purified, recombinant Human alpha Endosulfine (Accession#: O43768-1; Met1-Glu121). | |
| 100 μg | |
| Neuroscience | |
| 2029 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction