missing translation for 'onlineSavingsMsg'
Learn More

alpha-Endosulfan, TRC

Product Code. 30385921
Change view
Click to view available options
Quantity:
100 mg
2.5 g
Unit Size:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385921 2.5 g 2.5g
30386840 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30385921 Supplier Toronto Research Chemicals Supplier No. TRCE5555102.5G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 959-98-8
Molecular Formula C9 H6 Cl6 O3 S
Molecular Weight (g/mol) 406.93
Synonym 6,9-Methano-2,4,3-benzodioxathiepin, 6,7,8,9,10,10-hexachloro-1,5,5a,6,9,9a-hexahydro-, 3-oxide, (3α,5aβ,6α,9α,9aβ)-, 5-Norbornene-2,3-dimethanol, 1,4,5,6,7,7-hexachloro-, cyclic sulfite, endo- (8CI), Endosulfan A (7CI), Endosulfan 1, Endosulfan I, Endosulphan I, Thiodan I, α-Benzoepin, α-ES, α-Endosulfan, α-Thiodan, β-Thionex
IUPAC Name (3α,5aβ,6α,9α,9aβ)-6,7,8,9,10,10-hexachloro-1,5,5a,6,9,9a-hexahydro-6,9-methano-2,4,3-benzodioxathiepin 3-oxide
SMILES ClC1=C(Cl)[C@@]2(Cl)[C@H]3CO[S@@](=O)OC[C@H]3[C@]1(Cl)C2(Cl)Cl

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C9H6Cl6O3S/c10-5-6(11)8(13)4-2-18-19(16)17-1-3(4)7(5,12)9(8,14)15/h3-4H,1-2H2/t3-,4+,7-,8+,19+
Formula Weight 403.8169
Percent Purity >95%
Chemical Name or Material alpha-Endosulfan
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.