missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ alpha-ENaC Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA535364
This item is not returnable.
View return policy
Description
alpha-ENaC Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Epithelial sodium channels are amiloride-sensitive members of the degenerin/epithelial sodium channel (Deg/ENaC) superfamily of ion channels. Members of this superfamily of ion channels share organizational similarity in that they all possess two short intracellular amino and carboxyl termini, two short membrane spanning segments, and a large extracellular loop with a conserved cysteine-rich region. There are three homologous isoforms of the ENaC (alpha, beta, and gamma) protein. ENaC in the kidney, lung, and colon plays an essential role in trans-epithelial sodium and fluid balance. ENaC also mediates aldosterone-dependent sodium reabsorption in the distal nephron of the kidney, thus regulating blood pressure. ENaC is thought to be regulated, in part, through association with the cystic fibrosis transmembrane conductance regulator (CFTR) chloride ion channel. Gain-of-function mutations in beta- or gamma-ENaC can cause severe arterial hypertension (Liddel's syndrome) and loss-of-function mutations in alpha- or beta-ENaC causes pseudohypoaldosteronism (PHA-1).
Specifications
| alpha-ENaC | |
| Polyclonal | |
| Unconjugated | |
| SCNN1A | |
| alpha ENaC-2; alpha-ENaC; Alpha-NaCH; amiloride-sensitive epithelial sodium channel; amiloride-sensitive epithelial sodium channel alpha subunit; amiloride-sensitive sodium channel subunit alpha; amiloride-sensitive sodium channel subunit alpha 2; BESC2; ENaC; ENaC alpha; ENaCa; ENaCalpha; Epithelial Na(+) channel subunit alpha; epithelial sodium channel alpha subunit; FLJ21883; mENaC; nasal epithelial sodium channel alpha subunit; nonvoltage-gated sodium channel 1 subunit alpha; Renac; SCNEA; SCNN1; Scnn1a; sodium channel epithelial 1 alpha subunit; sodium channel, non voltage gated 1 alpha subunit; sodium channel, nonvoltage-gated 1 alpha; sodium channel, non-voltage-gated 1 alpha subunit; sodium channel, nonvoltage-gated, type I, alpha; sodium channel, nonvoltage-gated, type I, alpha polypeptide | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 6337 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P37088 | |
| SCNN1A | |
| KLH conjugated synthetic peptide between 365-391 amino acids from the central region of human SCNN1A. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction