missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ alpha Dystroglycan Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA519323
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine and canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
Dystroglycan is a laminin binding component of the dystrophin-glycoprotein complex which provides a linkage between the subsarcolemmal cytoskeleton and the extracellular matrix. Dystroglycan 1 is a candidate gene for the site of the mutation in autosomal recessive muscular dystrophies. The dramatic reduction of dystroglycan 1 in Duchenne muscular dystrophy leads to a loss of linkage between the sarcolemma and extracellular matrix, rendering muscle fibers more susceptible to necrosis. Dystroglycan also functions as dual receptor for agrin and laminin-2 in the Schwann cell membrane. The muscle and nonmuscle isoforms of dystroglycan differ by carbohydrate moieties but not protein sequence. Alternative splicing results in multiple transcript variants all encoding the same protein.
Specifications
| alpha Dystroglycan | |
| Polyclonal | |
| Unconjugated | |
| DAG1 | |
| 156DAG; A3a; AGRNR; Alpha-DG; Alpha-dystroglycan; Beta-DG; Beta-dystroglycan; D9Wsu13e; DAG; DAG1; Dag-1; DG; Dp427; Dp71; dystroglycan; dystroglycan 1; dystroglycan 1 (dystrophin-associated glycoprotein 1); dystrophin associated glycoprotein 1; Dystrophin-associated glycoprotein 1; MDDGA9; MDDGC7; MDDGC9 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 1605 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q14118 | |
| DAG1 | |
| Synthetic peptide sequence (HVGKHEYFMHATDK) corresponding to the internal amino acids of DAG1 (aa 569-582). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction