missing translation for 'onlineSavingsMsg'
Learn More

alpha-Defensin 1 Antibody (1B20), Novus Biologicals™

Product Code. 18330878 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18330878 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18330878 Supplier Novus Biologicals Supplier No. H00001667M03100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

alpha-Defensin 1 Monoclonal antibody specifically detects alpha-Defensin 1 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha-Defensin 1
Applications ELISA
Classification Monoclonal
Clone 1B20
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH27917
Gene Alias alpha 2, DEF1DEFA1B, DEFA2, defensin, alpha 1, myeloid-related sequence, defensin, alpha 1HP-1, HNP-1HP1, MRSMGC138393, myeloid-related sequence, neutrophil defensin 1
Host Species Mouse
Immunogen DEFA1 (AAH27917, 1 a.a. ~ 94 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MRTLAILAAILLVALQAQAEPLQARADEVAAAPEQIAADIPEVVVSLAWDESLAPKHPGSRKSMDCYCRIPACIAGERRYGTCIYQGRLWAFCC
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1667
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.