missing translation for 'onlineSavingsMsg'
Learn More

Alpha-Damascone, TRC

Product Code. 30382551
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382551 500 mg 500mg
30375177 250 mg -
30377588 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30382551 Supplier Toronto Research Chemicals Supplier No. TRCD627985500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 43052-87-5
Molecular Formula C13H20O
Molecular Weight (g/mol) 192.3
Synonym α-Damascone
IUPAC Name 1-(2,6,6-trimethylcyclohex-2-en-1-yl)but-2-en-1-one
SMILES CC=CC(=O)C1C(=CCCC1(C)C)C

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C13H20O/c1-5-7-11(14)12-10(2)8-6-9-13(12,3)4/h5,7-8,12H,6,9H2,1-4H3/b7-5+
Formula Weight 192.151
Chemical Name or Material Alpha-Damascone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.