missing translation for 'onlineSavingsMsg'
Learn More

alpha-D-(+)-Talose, 97%, Thermo Scientific Chemicals

Catalog Number 11454737
Click to view available options
Quantity:
0.1 g
0.5 g
This item is not returnable. View return policy
2595-98-4
C6H12O6
180.16
MFCD00135834
GZCGUPFRVQAUEE-UHFFFAOYNA-N
d-+-talose, aldehydo-d-talose, 2s,3s,4s,5r-2,3,4,5,6-pentahydroxyhexanal
99459
CHEBI:68461
2,3,4,5,6-pentahydroxyhexanal
OCC(O)C(O)C(O)C(O)C=O
This item is not returnable. View return policy
2595-98-4
C6H12O6
180.16
MFCD00135834
GZCGUPFRVQAUEE-UHFFFAOYNA-N
d-+-talose, aldehydo-d-talose, 2s,3s,4s,5r-2,3,4,5,6-pentahydroxyhexanal
99459
CHEBI:68461
2,3,4,5,6-pentahydroxyhexanal
OCC(O)C(O)C(O)C(O)C=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 133°C to 135°C
Quantity 0.1 g
Formula Weight 180.16
Percent Purity 97%
Chemical Name or Material alpha-D-(+)-Talose

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'