missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals alpha-D(+)-Glucose, 99+%, anhydrous
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 kg
2.5 kg
25 kg
Unit Size:
1kg
2.5kg
25kg
Beskrivning
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifikationer
Specifikationer
| Acidity | 0.15mL 0.1N NaOH/6g max. |
| CAS Min % | 99.0 |
| CAS Max % | 100.0 |
| Melting Point | 156.0°C to 158.0°C |
| Color | White |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99% min. (ELSD) (HPLC) |
| Packaging | Plastic bottle |
| Quantity | 1 kg |
| Beilstein | 31, 83 |
| Visa mer |
RUO – Research Use Only
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering