missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Cyano-o-tolunitrile, 80%, Thermo Scientific™
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Specifications
CAS Min % | 75.0 |
CAS Max % | 100.0 |
Melting Point | 76°C |
Color | Beige to Brown |
Infrared Spectrum | Authentic |
Assay Percent Range | 75% min. (GC) |
Packaging | Glass bottle |
Linear Formula | NCCH2C6H4CN |
Quantity | 25 g |
Beilstein | 09, 859 |
Show More |
Safety and Handling
Product Identifier
- a-Cyano-o-tolunitrile
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 3
- H330-Fatal if inhaled.
- H311-Toxic in contact with skin.
- H301-Toxic if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'