missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ alpha COP Recombinant Rabbit Monoclonal Antibody (23GB3485), Invitrogen™
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£162.00 - £387.00

Specifications

Antigen alpha COP
Clone 23GB3485
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30089812
GREENER_CHOICE
View Documents
Invitrogen™
MA552568
100 μL
£387.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30089809
View Documents
Invitrogen™
MA552569
20 μL
£162.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

alpha COP Recombinant Monoclonal antibody specifically detects alpha COP in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot
Specifications

Specifications

alpha COP
Western Blot, Immunocytochemistry
Unconjugated
Rabbit
Human, Mouse, Rat
P53621, Q8CIE6
12847, 1314, 304978
IgG
Affinity chromatography
Antibody
23GB3485
Recombinant Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
AILJK; alpha coat protein; alpha-coat protein; Alpha-COP; AU040324; coatomer protein complex subunit alpha; coatomer subunit alpha; COPA; HEPCOP; HEP-COP; proxenin; xenin; Xenopsin-related peptide
A synthetic peptide of human COPA (1-29AA).
Primary
-20°C
COPA
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.