missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ alpha COP Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142630
This item is not returnable.
View return policy
Beskrivelse
Additional Information: This antibody is expected to recognize both reported isoforms (NP_001091868.1; NP_004362.2). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:1,000.
In eukaryotic cells, protein transport between the endoplasmic reticulum and Golgi compartments is mediated in part by non-clathrin-coated vesicular coat proteins (COPs). Seven coat proteins have been identified, and they represent subunits of a complex known as coatomer. The subunits are designated alpha-COP, beta-COP, beta-prime-COP, gamma-COP, delta-COP, epsilon-COP, and zeta-COP. The alpha-COP, encoded by COPA, shares high sequence similarity with RET1P, the alpha subunit of the coatomer complex in yeast. Also, the N-terminal 25 amino acids of alpha-COP encode the bioactive peptide, xenin, which stimulates exocrine pancreatic secretion and may act as a gastrointestinal hormone. Alternative splicing results in multiple splice forms encoding distinct isoforms.
Tekniske data
| alpha COP | |
| Polyclonal | |
| Unconjugated | |
| COPA | |
| AILJK; alpha coat protein; alpha-coat protein; Alpha-COP; AU040324; coatomer protein complex subunit alpha; coatomer subunit alpha; COPA; HEPCOP; HEP-COP; proxenin; xenin; Xenopsin-related peptide | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 1314 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P53621 | |
| COPA | |
| Peptide with sequence CTRASNLENST. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion