missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ alpha COP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106967
This item is not returnable.
View return policy
Description
Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
In eukaryotic cells, protein transport between the endoplasmic reticulum and Golgi compartments is mediated in part by non-clathrin-coated vesicular coat proteins (COPs). Seven coat proteins have been identified, and they represent subunits of a complex known as coatomer. The subunits are designated alpha-COP, beta-COP, beta-prime-COP, gamma-COP, delta-COP, epsilon-COP, and zeta-COP. The alpha-COP, encoded by COPA, shares high sequence similarity with RET1P, the alpha subunit of the coatomer complex in yeast. Also, the N-terminal 25 amino acids of alpha-COP encode the bioactive peptide, xenin, which stimulates exocrine pancreatic secretion and may act as a gastrointestinal hormone. Alternative splicing results in multiple splice forms encoding distinct isoforms.
Specifications
| alpha COP | |
| Polyclonal | |
| Unconjugated | |
| COPA | |
| AILJK; alpha coat protein; alpha-coat protein; Alpha-COP; AU040324; coatomer protein complex subunit alpha; coatomer subunit alpha; COPA; HEPCOP; HEP-COP; proxenin; xenin; Xenopsin-related peptide | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1314 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| P53621 | |
| COPA | |
| Synthetic peptide directed towards the middle region of human COPA (aa 396-446). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction