missing translation for 'onlineSavingsMsg'
Learn More

Alpha-Chloroethyl Chloroformate, TRC

Product Code. 30011875
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011875 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30011875 Supplier Toronto Research Chemicals Supplier No. TRCC36606010G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50893-53-3
Molecular Formula C3H4Cl2O2
Molecular Weight (g/mol) 142.97
Synonym 1-Chlorocarbonyloxy-1-chloroethane, 1-Chloroethyl Carbonochloridate, 1-Chloroethyl Chloroformate, Chloroformic Acid 1-Chloroethyl Ester, alpha-Chloroethoxycarbonyl Chloride
IUPAC Name 1-chloroethyl carbonochloridate
SMILES CC(Cl)OC(=O)Cl

Specifications

Quantity 10 g
InChI Formula InChI=1S/C3H4Cl2O2/c1-2(4)7-3(5)6/h2H,1H3
Formula Weight 141.9588
Chemical Name or Material alpha-Chloroethyl Chloroformate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.