missing translation for 'onlineSavingsMsg'
Learn More

alpha-Chloro-o-xylene, 99%, Thermo Scientific Chemicals

Product Code. 10677442
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 552-45-4
Molecular Formula C8H9Cl
Molecular Weight (g/mol) 140.61
MDL Number MFCD00000899
InChI Key VQRBXYBBGHOGFT-UHFFFAOYSA-N
Synonym 2-methylbenzyl chloride, 1-chloromethyl-2-methylbenzene, o-xylyl chloride, alpha-chloro-o-xylene, o-methylbenzyl chloride, o-chloromethyl toluene, benzene, 1-chloromethyl-2-methyl, 2-methylbenzylchloride, 2-chloromethyl toluene, unii-eqt3v044tw
PubChem CID 11093
IUPAC Name 1-(chloromethyl)-2-methylbenzene
SMILES CC1=CC=CC=C1CCl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -2.0°C
Color Colorless to Yellow
Density 1.0800g/mL
Boiling Point 197.0°C to 199.0°C
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4CH2Cl
Refractive Index 1.5405 to 1.5425
Quantity 100 mL
Beilstein 05,364
Merck Index 15,10289
Specific Gravity 1.08
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,ketones and,chlorinated hydrocarbons
Formula Weight 140.61
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material α-Chloro-o-xylene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • a-Chloro-o-xylene
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'