missing translation for 'onlineSavingsMsg'
Learn More

alpha-Chloralose, MP Biomedicals™

Product Code. 11443140 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity unitSize
11443140 10 g 10g
1 options
This item is not returnable. View return policy

Product Code. 11443140

Brand: MP Biomedicals 0210045910

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as an anesthetic for laboratory animals

TRUSTED_SUSTAINABILITY

Specifications

CAS 15879-93-3
Quantity 10 g
Molecular Formula C8H11Cl3O6
MDL Number MFCD00005542
Synonym a-chloralose, 1-2r,5r,6s,6ar-6-hydroxy-2-trichloromethyl-3a,5,6,6a-tetrahydrofuro 2,3-d 1,3 dioxol-5-yl ethane-1,2-diol, r-1-2r,3ar,5r,6s,6ar-6-hydroxy-2-trichloromethyl tetrahydrofuro 2,3-d 1,3 dioxol-5-yl ethane-1,2-diol
InChI Key OJYGBLRPYBAHRT-OPKHMCHVSA-N
SMILES OCC(O)[C@H]1OC2O[C@@H](O[C@@H]2[C@H]1O)C(Cl)(Cl)Cl
IUPAC Name 1-[(2R,5R,6S,6aR)-6-hydroxy-2-(trichloromethyl)-tetrahydro-2H-furo[2,3-d][1,3]dioxol-5-yl]ethane-1,2-diol
Molecular Weight (g/mol) 309.52
PubChem CID 7057995
Formula Weight 309.53
Chemical Name or Material α-Chloralose
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.