missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ alpha-Cardiac Actin Monoclonal Antibody (C302), Invitrogen™

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA551025
This item is not returnable.
View return policy
Description
ACTC1 are part of actins, which are highly conserved proteins that are involved in various types of cell motility. Polymerization of globular actin (G-actin) leads to a structural filament (F-actin) in the form of a two-stranded helix. Each actin can bind to four others. The protein encoded by this gene belongs to the actin family which is comprised of three main groups of actin isoforms, alpha, beta, and gamma. The alpha actins are found in muscle tissues and are a major constituent of the contractile apparatus. Defects in this gene have been associated with idiopathic dilated cardiomyopathy (IDC) and familial hypertrophic cardiomyopathy (FHC).
Specifications
| alpha-Cardiac Actin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| P68032, P68035 | |
| ACTC1 | |
| Recombinant Human Anti-ACTC1 protein, Asp156-Gly368 (Accession #P68032). | |
| 200 μL | |
| Primary | |
| Human, Pig, Rat | |
| Antibody | |
| IgG2b κ |
| Immunohistochemistry (Paraffin), Western Blot | |
| C302 | |
| Unconjugated | |
| ACTC1 | |
| ACTC; Actc1; Actc-1; actin alpha cardiac; actin alpha cardiac 1; actin alpha cardiac muscle 1; actin, alpha cardiac muscle 1; Actin, alpha cardiac muscle 1, intermediate form; actin, alpha, cardiac 1; actin, alpha, cardiac muscle; actin, alpha, cardiac muscle 1; alpha-actin cardiac; alphac-actin; alpha-cardiac actin; ASD5; B-actin; cardiac muscle alpha actin 1; CMD1R; CMH11; LVNC4 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 100152267, 29275, 70 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction