missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Bromostyrene (~90%, ~1% Hydroquinone as stabilizer), TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | alpha-Bromostyrene |
| CAS | 98-81-7 |
| Molecular Formula | C8 H7 Br |
| Quantity | 1 g |
| Synonym | Styrene,, (1-Bromoethenyl)benzene, (1-Bromovinyl)benzene, 1-Bromo-1-phenylethene, 1-Bromo-1-phenylethylene, 1-Bromostyrene, 1-Phenylvinyl bromide, α-Bromostyrene |
| InChI Formula | InChI=1S/C8H7Br/c1-7(9)8-5-3-2-4-6-8/h2-6H,1H2 |
| SMILES | BrC(=C)c1ccccc1 |
| IUPAC Name | 1-bromoethenylbenzene |
| Molecular Weight (g/mol) | 183.05 |
| Formula Weight | 181.9731 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction