missing translation for 'onlineSavingsMsg'
Learn More

alpha-Bromo-2-chlorophenylacetic acid, 98%, Thermo Scientific™

Product Code. 11467196
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11467196 25 g 25g
11457196 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11467196 Supplier Thermo Scientific Alfa Aesar Supplier No. B25518.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29270-30-2
Molecular Formula C8H6BrClO2
Molecular Weight (g/mol) 249.488
MDL Number MFCD03428513
InChI Key XHAPROULWZYBGA-UHFFFAOYSA-N
Synonym alpha-bromo-2-chlorophenylacetic acid, 2-bromo-2-2-chlorophenyl acetic acid, 2-bromo-2-2'-chlorophenyl acetic acid, a-bromo 2-chlorophenylacetic acid, alpha-bromo-2-chlorophenyl acetic acid, alpha-bromo 2-chloro phenylacetic acid, bromo 2-chlorophenyl acetic acid, a-bromo-2-chlorophenylacetic acid, alapha-bromo-2-chloro-phenylacetic acid, benzeneacetic acid, alpha-bromo-2-chloro
PubChem CID 2774031
IUPAC Name 2-bromo-2-(2-chlorophenyl)acetic acid
SMILES C1=CC=C(C(=C1)C(C(=O)O)Br)Cl

Specifications

Melting Point 105°C to 108°C
Quantity 25 g
UN Number UN3261
Formula Weight 249.49
Percent Purity 98%
Chemical Name or Material alpha-Bromo-2-chlorophenylacetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.