missing translation for 'onlineSavingsMsg'
Learn More

alpha-Bromo-2-chlorophenylacetic acid, 98%, Thermo Scientific™

Product Code. 11457196
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11457196 5 g 5g
11467196 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11457196 Supplier Thermo Scientific Alfa Aesar Supplier No. B25518.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29270-30-2
Molecular Formula C8H6BrClO2
Molecular Weight (g/mol) 249.488
MDL Number MFCD03428513
InChI Key XHAPROULWZYBGA-UHFFFAOYSA-N
Synonym alpha-bromo-2-chlorophenylacetic acid, 2-bromo-2-2-chlorophenyl acetic acid, 2-bromo-2-2'-chlorophenyl acetic acid, a-bromo 2-chlorophenylacetic acid, alpha-bromo-2-chlorophenyl acetic acid, alpha-bromo 2-chloro phenylacetic acid, bromo 2-chlorophenyl acetic acid, a-bromo-2-chlorophenylacetic acid, alapha-bromo-2-chloro-phenylacetic acid, benzeneacetic acid, alpha-bromo-2-chloro
PubChem CID 2774031
IUPAC Name 2-bromo-2-(2-chlorophenyl)acetic acid
SMILES C1=CC=C(C(=C1)C(C(=O)O)Br)Cl

Spezifikation

Melting Point 105°C to 108°C
Quantity 5 g
UN Number UN3261
Formula Weight 249.49
Percent Purity 98%
Chemical Name or Material alpha-Bromo-2-chlorophenylacetic acid
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.