missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-BHC, SPEX CertiPrep™
Shop All SPEX Certiprep Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
| Analyte | CAS No. | Concentration |
|---|---|---|
| alpha-BHC | 319-84-6 | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
| Packaging | 2mL amber prescored glass ampule |
| CAS | 319-84-6, 67-56-1 |
| CAS Min % | 0.1, 99.9 |
| Concentration or Composition Notes | Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. |
| CAS Max % | 0.1, 99.9 |
| Physical Form | Solid |
| Molecular Formula | C6H6Cl6 |
| Color | White |
| pH | 7.5 to 9.5 |
| Melting Point | 300°C |
| Show More |
Safety and Handling
Product Identifier
- alpha-BHC
- Danger
- Acute toxicity Category 3
- Carcinogenicity Category 2
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H331-Toxic if inhaled.
- H351-Suspected of causing cancer.
- H370-Causes damage to organs.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : (1alpha,2alpha,3ß,4alpha,5ß,6ß)-1,2,3,4,5,6-hexachlorocyclohexane, methanol
Product intended for laboratory use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction