missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha B Crystallin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15808-1-AP-150UL
This item is not returnable.
View return policy
Description
Lens proteins consist almost entirely of crystallins (about 95%). Crystallins are also found vertebrate skeletal muscle tissue. In the lens, their structural function is to assist in maintaining the proper refractive index of the lens. The mammalian lens contains 3 major classes of crystallins: alpha, beta, and gamma. Alpha-crystallin is the largest of the crystallins and is composed of 2 primary gene products-alpha-A and alpha-B. There are at least 5 different proteins comprising the beta-crystallins. The gamma-crystallins are monomeric, but there are at least 5 gamma crystallins identified in bovine and rat lens. Alpha-Crystallin comprises 40% of total lens protein composition. In addition to maintaining proper refractive index, it also functions in a chaperone like manner by preventing the formation of aggregates possibly leading to cataract formation. It is believed that the phosphorylated states of the alpha-crystallin occur in response to cellular stress and may serve a structural control function and play a role in protein maintece. Alpha-B crystallin has been linked to Alexander′s disease where it accumulates in brain cells of those afflicted.Specifications
| Alpha B Crystallin | |
| Polyclonal | |
| Unconjugated | |
| CRYAB | |
| Alpha B Crystallin, Alpha crystallin B chain, Alpha(B) crystallin, CRYA2, CRYAB, crystallin, alpha B, CTPP2, Heat shock protein beta 5, HSPB5, Rosenthal fiber component | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12955, 1410, 25420 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P02511, P23927, P23928 | |
| Cryab | |
| Alpha B Crystallin Fusion Protein Ag8543 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction