missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha B Crystallin Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68001-1-IG-150UL
This item is not returnable.
View return policy
Description
The Alpha B Crystallin antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Alpha B Crystallin. This antibody recognizes human, mouse, rat antigen. The Alpha B Crystallin antibody has been validated for the following applications: WB, IHC, ELISA analysis.Specifications
| Alpha B Crystallin | |
| Monoclonal | |
| 1000 μg/mL | |
| CRYA2; CTPP2; HSPB5 | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 20-22 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, ELISA | |
| 1B4C3 | |
| Unconjugated | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 1410 | |
| Store at -20°C. | |
| Monoclonal Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction