missing translation for 'onlineSavingsMsg'
Learn More

alpha-Asarone

Product Code. p-7043025
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15466739

Brand: Thermo Scientific Alfa Aesar J65125.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2883-98-9 | C12H16O3 | 208.257 g/mol

α-Asarone was used in the synthesis of series of α-asarone isomers and were investigated for their hypolipidemic and antiplatelet activities. It is used in the synthesis of anti-inflammatory neolignan. Also used in lignan syntheses for antiallergic agents. It is clinically used as medication for treating epilepsy, cough, bronchitis and asthma. It exhibits neuroprotective, anti-oxidative, anticonvulsive and cognitive enhancing action.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in chloroform, methanol.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat. Store in refrigerator.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2883-98-9
Molecular Formula C12H16O3
Molecular Weight (g/mol) 208.257
MDL Number MFCD00064457
InChI Key RKFAZBXYICVSKP-AATRIKPKSA-N
Synonym alpha-asarone, asarone, trans-isoasarone, trans-asarone, asaron, azaron, etherophenol, trans-isoasaron, e-asarone, asarum camphor
PubChem CID 636822
ChEBI CHEBI:78309
IUPAC Name 1,2,4-trimethoxy-5-[(E)-prop-1-enyl]benzene
SMILES CC=CC1=CC(=C(C=C1OC)OC)OC

Specifications

Melting Point 57°C to 61°C
Color White to Yellow
Boiling Point 296°C
Linear Formula (CH3O)3C6H2CH=CHCH3
Quantity 1 g
Beilstein 1910606
Sensitivity Light sensitive
Merck Index 14,824
Solubility Information Soluble in chloroform,methanol.
Formula Weight 208.25
Physical Form Powder
Chemical Name or Material alpha-Asarone
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.