missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ alpha Amylase 1 Polyclonal Antibody
Sheep Polyclonal Antibody
Brand: Invitrogen™ PA185176
This item is not returnable.
View return policy
Description
PA1-85176 also recognizes porcine pancreatic amylase (50%) and weakly fungal amylase (10%).
Amylase is an enzyme that catalyses the breakdown of starch into sugars. Amylase is present in human saliva, where it begins the chemical process of digestion. By in situ hybridization combined with high resolution cytogenetics, the amylase gene is mapped to 1p21. Amylase enzymes find use in bread making and to break down complex sugars such as starch (found in flour) into simple sugars. Yeast then feeds on these simple sugars and converts it into the waste products of alcohol and CO2.
Specifications
| alpha Amylase 1 | |
| Polyclonal | |
| Unconjugated | |
| AMY1A | |
| 1,4-alpha-D-glucan glucanohydrolase 1; alpha amylase 1; alpha-amylase 1; Amy1; Amy-1; Amy1a; Amy-1-a; AMY1B; AMY1C; AMY2A; amylase 1, salivary; amylase, alpha 1A (salivary); amylase, salivary, alpha-1A; C030014B17Rik; glycogenase; PA; salivary alpha-amylase; salivary amylase; salivary amylase alpha 1A; salivary and hepatic alpha-amylase | |
| Sheep | |
| Affinity chromatography | |
| RUO | |
| 276 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Frozen) | |
| 5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P0DUB6 | |
| AMY1A | |
| highly pure human salivary amylase. | |
| 1 mL | |
| Primary | |
| Fungi, Human, Pig | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction