missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ alpha Amylase 1 Monoclonal Antibody (C3), Invitrogen™

Mouse Monoclonal Antibody
£161.00 - £361.00
Specifications
| Antigen | alpha Amylase 1 |
|---|---|
| Clone | C3 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17920783
|
Invitrogen™
MA551016 |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17989852
![]() |
Invitrogen™
MA551015 |
200 μL |
£361.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Amylase is an enzyme that catalyses the breakdown of starch into sugars. Amylase is present in human saliva, where it begins the chemical process of digestion. By in situ hybridization combined with high resolution cytogenetics, the amylase gene is mapped to 1p21. Amylase enzymes find use in bread making and to break down complex sugars such as starch (found in flour) into simple sugars. Yeast then feeds on these simple sugars and converts it into the waste products of alcohol and CO2.Specifications
| alpha Amylase 1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| 1,4-alpha-D-glucan glucanohydrolase 1; alpha amylase 1; alpha-amylase 1; Amy1; Amy-1; Amy1a; Amy-1-a; AMY1B; AMY1C; AMY2A; amylase 1, salivary; amylase, alpha 1A (salivary); amylase, salivary, alpha-1A; C030014B17Rik; glycogenase; PA; salivary alpha-amylase; salivary amylase; salivary amylase alpha 1A; salivary and hepatic alpha-amylase | |
| Amy1, AMY1A | |
| IgG1 κ | |
| Protein A/G | |
| AMY1A |
| C3 | |
| Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| P00687, P0DUB6 | |
| 11722, 24203, 276 | |
| Recombinant Human AMY1A protein, Ala15-Leu511 (Accession #P04745). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title