missing translation for 'onlineSavingsMsg'
Learn More

alpha-Aminoadipate Aminotransferase Antibody [Janelia Fluor™ 525], Novus Biologicals™

Product Code. 30734148 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30734148 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734148 Supplier Novus Biologicals Supplier No. AF7927JF525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

alpha-Aminoadipate Aminotransferase Polyclonal antibody specifically detects alpha-Aminoadipate Aminotransferase in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha-Aminoadipate Aminotransferase
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias AadAT, aminoadipate aminotransferase, EC 2.6.1.39,2-aminoadipate aminotransferase, EC 2.6.1.7,2-aminoadipate transaminase, KAT/AadAT, KAT2mitochondrial, Kynurenine aminotransferase II, Kynurenine--oxoglutarate aminotransferase II, Kynurenine--oxoglutarate transaminase II, L kynurenine/alpha aminoadipate aminotransferase
Host Species Sheep
Immunogen E. coli-derived recombinant human alpha -Aminoadipate Aminotransferase, Met1-Leu425, Accession # Q8N5Z0
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Neurodegeneration, Neuroscience, Vision
Primary or Secondary Primary
Gene ID (Entrez) 51166
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.