missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha-Aminoacetophenone (Mixed Salt, >85%), TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Alpha-Aminoacetophenone |
| CAS | 5468-37-1 |
| Molecular Formula | C8H9NO . xHCl . xHBr |
| Quantity | 5 g |
| Synonym | 2-Amino-1-phenylethanone, 2-Amino-1-phenyl-1-ethanone, 2-Amino-1-phenylethanone, 2-Aminoacetophenone, Phenacylamine, α-Aminoacetophenone |
| InChI Formula | InChI=1S/C8H9NO.ClH/c9-6-8(10)7-4-2-1-3-5-7;/h1-5H,6,9H2;1H |
| SMILES | Cl.NCC(=O)c1ccccc1 |
| IUPAC Name | 2-amino-1-phenylethanone;hydrochloride |
| Molecular Weight (g/mol) | 171.62 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction