missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-alpha-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid

Product Code. 15480397
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 15480397

Brand: Thermo Scientific Alfa Aesar J64884.MA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 74341-63-2 | C7H10N2O4 | 186.167 g/mol

Selective quisqualate agonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 74341-63-2
Color White
Molecular Formula C7H10N2O4
MDL Number MFCD00213388
Quantity 10 mg
Synonym ampa, r,s-ampa, ampa pharmaceutical, alpha-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid, rs-ampa, +/--ampa, chembl13378, d,l-alpha-amino-3-hydroxy-5-methylisoxazole-4-propionic acid, r,s-alpha-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid, 4-isoxazolepropanoic acid, alpha-amino-2,3-dihydro-5-methyl-3-oxo
Solubility Information Soluble in warm water at 1mg/ml; Also soluble in 0;1M NaOH or 0;1M HCl at approximately 4mg/ml; Soluble in DMSO at 4mg/ml
InChI Key UUDAMDVQRQNNHZ-UHFFFAOYSA-N
SMILES CC1=C(C(=O)NO1)CC(C(=O)O)N
IUPAC Name 2-amino-3-(5-methyl-3-oxo-1,2-oxazol-4-yl)propanoic acid
Molecular Weight (g/mol) 186.167
PubChem CID 1221
ChEBI CHEBI:34018
Formula Weight 186.17
Physical Form Powder
Chemical Name or Material (+/-)-alpha-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.