missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha,Alpha-[UL-D14]Trehalose, TRC
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Specifications
Specifications
| Chemical Name or Material | alpha,alpha-[UL-D14]Trehalose |
| Molecular Formula | C12D14H8O11 |
| Quantity | 10 mg |
| Synonym | 1-O-α-D-[UL-D7]Glucopyranosyl-α-D-[UL-D7]glucopyranoside |
| InChI Formula | InChI=1S/C12H22O11/c13-1-3-5(15)7(17)9(19)11(21-3)23-12-10(20)8(18)6(16)4(2-14)22-12/h3-20H,1-2H2/t3-,4-,5-,6-,7+,8+,9-,10-,11-,12-/m1/s1/i1D2,2D2,3D,4D,5D,6D,7D,8D,9D,10D,11D,12D |
| SMILES | O[C@]1([2H])[C@](O)([2H])[C@@](O)([2H])[C@](C([2H])([2H])O)([2H])O[C@]1([2H])O[C@]2([2H])[C@@](O)([2H])[C@](O)([2H])[C@@](O)([2H])[C@](C([2H])([2H])O)([2H])O2 |
| Molecular Weight (g/mol) | 356.39 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction