missing translation for 'onlineSavingsMsg'
Learn More

α,α,α-Trifluoroacetophenone, 99%, Thermo Scientific Chemicals

Product Code. 10648054
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 434-45-7
Molecular Formula C8H5F3O
Molecular Weight (g/mol) 174.12
MDL Number MFCD00000420
InChI Key KZJRKRQSDZGHEC-UHFFFAOYSA-N
Synonym trifluoroacetophenone, 2,2,2-trifluoroacetophenone, 2,2,2-trifluoro-1-phenylethan-1-one, phenyl trifluoromethyl ketone, trifluoromethyl phenyl ketone, alpha,alpha,alpha-trifluoroacetophenone, ethanone, 2,2,2-trifluoro-1-phenyl, 2,2,2-trifluoro-1-phenyl-ethanone, trifluoroacetylbenzene, unii-6t7l1upy09
PubChem CID 9905
IUPAC Name 2,2,2-trifluoro-1-phenylethanone
SMILES C1=CC=C(C=C1)C(=O)C(F)(F)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -40.0°C
Color Colorless to Yellow
Density 1.2400g/mL
Boiling Point 165.0°C to 166.0°C
Flash Point 41°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5COCF3
Refractive Index 1.4580 to 1.4600
Quantity 100 g
Fieser 01,1226; 03,308
Specific Gravity 1.24
Solubility Information Solubility in water: insoluble
Formula Weight 174.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material α, α, α-Trifluoroacetophenone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'