missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha adducin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10791-1-AP-150UL
This item is not returnable.
View return policy
Description
Adducins are a family of cytoskeleton proteins encoded by three genes (alpha, beta, gamma). Adducin is a heterodimeric protein that consists of related subunits, which are produced from distinct genes but share a similar structure. Alpha- and beta-adducin include a protease-resistant N-terminal region and a protease-sensitive, hydrophilic C-terminal region. Alpha- and gamma-adducins are ubiquitously expressed. In contrast, beta-adducin is expressed at high levels in brain and hematopoietic tissues. Adducin binds with high affinity to Ca(2+)/calmodulin and is a substrate for protein kinases A and C. Alternative splicing results in multiple variants encoding distinct isoforms; however, not all variants have been fully described.Specifications
| Alpha adducin | |
| Polyclonal | |
| Unconjugated | |
| ADD1 | |
| ADD1, ADDA, adducin 1 (alpha), Alpha adducin | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11518, 118, 24170 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P35611, Q63028, Q9QYC0 | |
| Add1 | |
| Alpha adducin Fusion Protein Ag1245 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction