missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ alpha Adducin Polyclonal Antibody

Rabbit Polyclonal Antibody
£178.00 - £430.00
Specifications
| Antigen | alpha Adducin |
|---|---|
| Concentration | 0.5 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17700354
![]() |
Invitrogen™
PA5118937 |
200 μL |
£430.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17703674
|
Invitrogen™
PA5122272 |
20 μL |
£178.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
alpha Adducin Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Adducins are a family of cytoskeleton proteins encoded by three genes (alpha, beta, gamma). Adducin is a heterodimeric protein that consists of related subunits, which are produced from distinct genes but share a similar structure. Alpha- and beta-adducin include a protease-resistant N-terminal region and a protease-sensitive, hydrophilic C-terminal region. Alpha- and gamma-adducins are ubiquitously expressed. In contrast, beta-adducin is expressed at high levels in brain and hematopoietic tissues. Adducin binds with high affinity to Ca(2+)/calmodulin and is a substrate for protein kinases A and C. Alternative splicing results in multiple variants encoding distinct isoforms; however, not all variants have been fully described.Specifications
| alpha Adducin | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse | |
| ADD1 | |
| Add1; ADD2; ADDA; ADDB; adducin 1; adducin 1 (alpha); Adducin beta; Adducin1; Adducin-1; Adducin-alpha; Adducin-beta; AI256389; Alpha adducin; alpha-ADD; alpha-adducin; beta Adducin; Beta-adducin; erythrocyte adducin alpha subunit; erythrocyte adducin subunit alpha | |
| ADD1 | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P35611, Q9QYC0 | |
| 11518, 118 | |
| Recombinant protein Alpha Adducin. The antigen corresponds to amino acid range 388-560 of the target protein. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title