missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ alpha Adducin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599523
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ADD1.
Adducins are a family of cytoskeleton proteins encoded by three genes (alpha, beta, gamma). Adducin is a heterodimeric protein that consists of related subunits, which are produced from distinct genes but share a similar structure. Alpha- and beta-adducin include a protease-resistant N-terminal region and a protease-sensitive, hydrophilic C-terminal region. Alpha- and gamma-adducins are ubiquitously expressed. In contrast, beta-adducin is expressed at high levels in brain and hematopoietic tissues. Adducin binds with high affinity to Ca(2+)/calmodulin and is a substrate for protein kinases A and C. Alternative splicing results in multiple variants encoding distinct isoforms; however, not all variants have been fully described.
Specifications
| alpha Adducin | |
| Polyclonal | |
| Unconjugated | |
| ADD1 | |
| Add1; ADD2; ADDA; ADDB; adducin 1; adducin 1 (alpha); Adducin beta; Adducin1; Adducin-1; Adducin-alpha; Adducin-beta; AI256389; Alpha adducin; alpha-ADD; alpha-adducin; beta Adducin; Beta-adducin; erythrocyte adducin alpha subunit; erythrocyte adducin subunit alpha | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11518, 118, 24170 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P35611, Q63028, Q9QYC0 | |
| ADD1 | |
| A synthesized peptide derived from human ADD1(Accession P35611), corresponding to amino acid residues G700-S737. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction