missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha Adducin Antibody (5D4H1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£435.00
Specifications
| Antigen | alpha Adducin |
|---|---|
| Clone | 5D4H1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF |
Description
alpha Adducin Monoclonal specifically detects alpha Adducin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| alpha Adducin | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| Unconjugated | |
| Mouse | |
| Phospho Specific | |
| PBS with 0.05% Sodium Azide | |
| 118 | |
| Purified recombinant fragment of human ADD1 (AA: 1-193) expressed in E. Coli. | |
| Primary | |
| 81 kDa |
| 5D4H1 | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ADDA, adducin 1 (alpha), alpha-adducin, erythrocyte adducin alpha subunit, Erythrocyte adducin subunit alpha, MGC3339, MGC44427 | |
| ADD1 | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title