missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha Adaptin Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92798-0.1ml
This item is not returnable.
View return policy
Description
alpha Adaptin Polyclonal antibody specifically detects alpha Adaptin in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| alpha Adaptin | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| 100 kDa coated vesicle protein A, Adapter-related protein complex 2 alpha-1 subunit, Adaptor protein complex AP-2 subunit alpha-1, adaptor-related protein complex 2, alpha 1 subunit, ADTAAAP2-ALPHA, alpha1-adaptin, Alpha-adaptin A, AP-2 complex subunit alpha-1, CLAPA1adaptin, alpha A, Clathrin assembly protein complex 2 alpha-A large chain, clathrin-associated/assembly/adaptor protein, large, alpha 1, Plasma membrane adaptor HA2/AP2 adaptin alpha A subunit | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human AP2A1 (NP_055018.2). MPAVSKGDGMRGLAVFISDIRNCKSKEAEIKRINKELANIRSKFKGDKALDGYSKKKYVCKLLFIFLLGHDIDFGHMEAVNLLSSNKYTEKQIGYLFISVLVNSNSELIRLINNAIKNDLASRNPTFMCLALHCIANVGSREMGEAFAADIPRILVAGDSMDSVKQSAALCLLRLYKASPDLVPMGEWTARVVHLLNDQHMGVVTAAVSLITCLCKKNPDDFKTCVSLAVSRLSRIVSSASTDLQDYTYYFVPAPWLSVK | |
| 0.1 mL | |
| Cell Biology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 160 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction