missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha Actinin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11313-2-AP-20UL
This item is not returnable.
View return policy
Description
Alpha Actinin 1 belong to the spectrin gene superfamily which represents a diverse group of cytoskeletal proteins, including the alpha and beta spectrins and dystrophins. Alpha actinin is an actin-binding protein with multiple roles in different cell types. In nonmuscle cells, the cytoskeletal isoform is found along microfilament bundles and adherens-type junctions, where it is involved in binding actin to the membrane. In contrast, skeletal, cardiac, and smooth muscle isoforms are localized to the Z-disc and analogous dense bodies, where they help anchor the myofibrillar actin filaments. This gene encodes a nonmuscle, cytoskeletal, alpha actinin isoform and maps to the same site as the structurally similar erythroid beta spectrin gene. Three transcript variants encoding different isoforms have been found for this gene.Specifications
| alpha Actinin | |
| Polyclonal | |
| Unconjugated | |
| ACTN1 | |
| actinin, alpha 1, ACTN1, alpha Actinin, Alpha actinin 1, F actin cross linking protein, Non muscle alpha actinin 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 109711, 81634, 87 | |
| -20°C | |
| Liquid |
| Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P12814, Q7TPR4, Q9Z1P2 | |
| ACTN1 | |
| alpha Actinin Fusion Protein Ag1859 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction