missing translation for 'onlineSavingsMsg'
Learn More

Alpha Actinin 3 Rabbit anti-Human, Mouse, Rat, Clone: 4H7E6, Novus Biologicals™

Product Code. 18391992 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18391992 20 μg 20µL
18365322 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18391992 Supplier Novus Biologicals Supplier No. NBP31582720UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Alpha Actinin 3 Monoclonal antibody specifically detects Alpha Actinin 3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Alpha Actinin 3
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 4H7E6
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias actinin, alpha 3, alpha-actinin skeletal muscle, Alpha-actinin skeletal muscle isoform 3, alpha-actinin-3, F-actin cross-linking protein, MGC117002, MGC117005
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 700-800 of human Alpha Actinin 3 (Q08043).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Cytoskeleton Markers, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 89
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.