Learn More
Invitrogen™ alpha Actinin 3 Monoclonal Antibody (9B5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA533007
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat skeletal muscle tissue, mouse skeletal muscle tissue. IHC: human skeletal muscle tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Alpha-actinin-3, also known as alpha-actinin skeletal muscle isoform 3 or F-actin cross-linking protein, is a protein that in humans is encoded by the ACTN3 gene. This gene encodes a member of the alpha-actin binding protein gene family. The encoded protein is primarily expressed in skeletal muscle and functions as a structural component of sarcomeric Z line. This protein is involved in crosslinking actin containing thin filaments. An allelic polymorphism in this gene results in both coding and non-coding variants; the reference genome represents the coding allele.
Specifications
| alpha Actinin 3 | |
| Monoclonal | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| O88990, Q08043 | |
| Actn3 | |
| A synthetic peptide corresponding to a sequence at the C-terminus of human ACTN3 (574-617aa EADRERGAIMGIQGEIQKICQTYGLRPCSTNPYITLSPQDINT K). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 9B5 | |
| Unconjugated | |
| Actn3 | |
| actinin alpha 3; actinin alpha 3 (gene/pseudogene); Actinin alpha3; actinin, alpha 3; Actinin-alpha3; ACTN3; alpha-actinin skeletal muscle; Alpha-actinin skeletal muscle isoform 3; alpha-Actinin3; Alpha-actinin-3; alphaActinin-3; F-actin cross-linking protein | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 11474, 171009, 89 | |
| -20°C | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.